Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Bactenecin
Bovine Bactenecin
Catalog # 103006-332
Supplier:  Anaspec Inc
CAS Number:  
Bovine Bactenecin
Catalog # 103006-332
Supplier:  Anaspec Inc
Supplier Number:  AS-61066
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Bovine
  • Size:
    0.5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Bactenacin antimicrobial
  • Protein/Peptide Name:
    Bactenecin
  • Purity:
    95%
  • Molecular Weight:
    1483.9 Da
  • Sequence:
    RLCRIVVIRVCR (Disulfide bridge: 3-11)
  • Cat. No.:
    103006-332
  • Supplier no.:
    AS-61066

Specifications

About this item

Bactenecin is a 12-amino acid cationic antimicrobial peptide from bovine neutrophils.
Sequence:RLCRIVVIRVCR (Disulfide bridge: 3-11)
MW:1483.9 Da
% peak area by HPLC:95
Storage condition:-20° C