Order Entry
Puerto Rico
ContactUsLinkComponent
Plasmodium Plasmepsin V FRET Substrate, DABCYL-EDANS
Plasmodium Plasmepsin V FRET Substrate, DABCYL-EDANS
Catalog # 103008-598
Supplier:  Anaspec
CAS Number:  
Plasmodium Plasmepsin V FRET Substrate, DABCYL-EDANS
Catalog # 103008-598
Supplier:  Anaspec
Supplier Number:  AS-64939
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    DABCYL-EDANS
  • Species:
    Plasmodium
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Plasmepsin Peptide|PMV substrate Peptide
  • Protein/Peptide Name:
    Plasmepsin V FRET Substrate
  • Purity:
    95%
  • Molecular Weight:
    1751.1 Da
  • Sequence:
    Dabcyl-LNKRLLHETQ-EDANS
  • Cat. No.:
    103008-598
  • Supplier no.:
    AS-64939

Specifications

About this item

This FRET substrate peptide for Plasmepsin V (PMV) is derived from the conserved Plasmodium Export Element (PEXEL) motif of Histidine-Rich Protein II (HRPII). PMV is an ER aspartic protease that recognizes and cleaves the RXL sequence within the PEXEL motif of proteins exported by human malaria parasite Plasmodium falciparum, allowing them to translocate into host erythrocytes.
Sequence:Dabcyl-LNKRLLHETQ-EDANS
MW:1751.1 Da
% peak area by HPLC:95
Storage condition:-20° C