Order Entry
Puerto Rico
ContactUsLinkComponent
Cls Substrate
Cls Substrate
Catalog # 103006-568
Supplier:  Anaspec Inc
CAS Number:  
Cls Substrate
Catalog # 103006-568
Supplier:  Anaspec Inc
Supplier Number:  AS-61314
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    0.5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    C2 FAM|Complement component Peptide|C2 Peptide|Cls Substrate Peptide
  • Protein/Peptide Name:
    Cls Substrate
  • Purity:
    95%
  • Molecular Weight:
    2019.2 Da
  • Sequence:
    5-FAM-SLGRKIQIK(QXL 520)-NH2
  • Cat. No.:
    103006-568
  • Supplier no.:
    AS-61314

Specifications

About this item

This peptide is a second complement component (C2), the physiological substrate for the proenzyme Cls, first complement component. The complement system is a central component of host defense but can also contribute to the inflammation seen in pathological conditions. The C1s protease of the C1 complex initiates the host defense pathway. This peptide employs 5FAM/QXL520 FRET pair for quantitation of complement enzyme activity.
Sequence:5-FAM-SLGRKIQIK(QXL 520)-NH2
MW:2019.2 Da
% peak area by HPLC:95
Storage condition:-20° C