Order Entry
Puerto Rico
ContactUsLinkComponent
Bactenecin 2A
Bactenecin 2A
Catalog # 103008-584
Supplier:  Anaspec Inc
CAS Number:  
Bactenecin 2A
Catalog # 103008-584
Supplier:  Anaspec Inc
Supplier Number:  AS-64906
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Bactenacin antimicrobial Peptide
  • Protein/Peptide Name:
    Bactenecin 2A
  • Purity:
    95%
  • Molecular Weight:
    1420.8 Da
  • Sequence:
    RLARIVVIRVAR-NH2
  • Cat. No.:
    103008-584
  • Supplier no.:
    AS-64906

Specifications

About this item

This peptide is Bac2A, a linear variant of the loop-shaped cationic antimicrobial peptide Bactenecin found in bovine neutrophils. The Cys disulfide bond-forming residues of Bactenecin is replaced with Ala in Bac2A, and is active against both gram-positive and gram-negative bacteria.
Sequence:RLARIVVIRVAR-NH2
MW:1420.8 Da
% peak area by HPLC:95
Storage condition:-20° C