Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:C3a anaphylatoxin Peptide
- Protein/Peptide Name:C3a (70-77)
- Purity:95%
- Molecular Weight:823.9 Da
- Sequence:ASHLGLAR
- Cat. No.:103006-356
- Supplier no.:AS-61118
This octapeptide is a COOH-terminal fragment of the C3a anaphylatoxin peptide. On a molar basis, this peptide possesses 1-2% of the biological activities of C3a. It causes contraction of rodent ileum and uterus, release of vasoactive amines from rat mast cells, and increases vascular permeability in guinea pig and human skin. Both purified C3a and synthetic C3a (70-77), which retains partially the activity of anaphylatoxin, are shown to interact directly with human lymphocytes.
Sequence:ASHLGLAR
MW:823.9 Da
% peak area by HPLC:95
Storage condition:-20° C