Order Entry
Puerto Rico
ContactUsLinkComponent
MBP (84-97)
MBP (84-97)
Catalog # 103007-492
Supplier:  Anaspec Inc
CAS Number:  
MBP (84-97)
Catalog # 103007-492
Supplier:  Anaspec Inc
Supplier Number:  AS-62731
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Myelin basic protein 84-97
  • Protein/Peptide Name:
    MBP (84-97)
  • Purity:
    95%
  • Molecular Weight:
    1655 Da
  • Sequence:
    VVHFFKNIVTPRTP
  • Cat. No.:
    103007-492
  • Supplier no.:
    AS-62731

Specifications

About this item

This is fragment of the myelin basic protein (MBP), which induces moderate experimental autoimmune encephalomyelitis (EAE) symptoms in immunized mice. It corresponds to amino acids 83-96 of the murine sequence (85-98 in guinea pig; 86-99 in human).
Sequence:VVHFFKNIVTPRTP
MW:1655 Da
% peak area by HPLC:95
Storage condition:-20° C