Order Entry
Puerto Rico
ContactUsLinkComponent
Integrin-Binding Site
Integrin-Binding Site
Catalog # 103007-154
Supplier:  Anaspec Inc
CAS Number:  
Integrin-Binding Site
Catalog # 103007-154
Supplier:  Anaspec Inc
Supplier Number:  AS-62049
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    GRGDNP Peptide|Integrin-Binding sequence
  • Protein/Peptide Name:
    Integrin-Binding Site
  • Purity:
    95%
  • Molecular Weight:
    614.6 Da
  • Sequence:
    GRGDNP
  • Cat. No.:
    103007-154
  • Supplier no.:
    AS-62049

Specifications

About this item

This RGD-containing sequence is integrin-binding site. RGD peptides are found in both extracellular matrix (ECM) (e.g., collagen, osteopontin, fibronectin, and vitronectin) and non-ECM proteins (e.g., disintegrins). RGD-containing peptides are recognized by several integrins. This peptide targets both alphavbeta3 and alpha5beta1 integrins. Interaction of the RGDN sequence with endothelial alpha5beta1 integrin causes endothelin-mediated arteriolar vasoconstriction. This peptide influences the IkappaB kinase (IKK)/nuclear factor-kappaB (NF-kappaB) activation. It also controls the cardiac contractile function.
Sequence:GRGDNP
MW:614.6 Da
% peak area by HPLC:95
Storage condition:-20° C