Order Entry
Puerto Rico
ContactUsLinkComponent
Cyclo (RGDfC) Peptide
Cyclo (RGDfC) Peptide
Catalog # 103007-726
Supplier:  Anaspec
CAS Number:  
Cyclo (RGDfC) Peptide
Catalog # 103007-726
Supplier:  Anaspec
Supplier Number:  AS-63785-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    avb3 Integrin binding Peptide|RGD Peptide|RGDfC Peptide
  • Protein/Peptide Name:
    Cyclo (RGDfC) Peptide
  • Purity:
    95%
  • Molecular Weight:
    578.7 Da
  • Sequence:
    Cyclo(-RGDfC)
  • Cat. No.:
    103007-726
  • Supplier no.:
    AS-63785-1

Specifications

About this item

This is a cyclic RGDfC sequence, an integrin avb3-affinity peptide.
Sequence:Cyclo(-RGDfC)
MW:578.7 Da
% peak area by HPLC:95
Storage condition:-20° C