Order Entry
Puerto Rico
ContactUsLinkComponent
[Glu3]-RGES, Control for RGD Peptides
[Glu3]-RGES, Control for RGD Peptides
Catalog # 103007-382
Supplier:  Anaspec Inc
CAS Number:  
[Glu3]-RGES, Control for RGD Peptides
Catalog # 103007-382
Supplier:  Anaspec Inc
Supplier Number:  AS-62527
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    RGES control Peptide|Control for RGDS Fibronectin Active Fragment|RGD control Peptide
  • Protein/Peptide Name:
    [Glu3]-RGES, Control for RGD Peptides
  • Purity:
    95%
  • Molecular Weight:
    447.5 Da
  • Sequence:
    RGES
  • Cat. No.:
    103007-382
  • Supplier no.:
    AS-62527

Specifications

About this item

This peptide is a control for the RGDS Fibronectin Active Fragment and other RGD-related peptides. Asp3 is replaced by Glu3 in RGDS peptide changing its properties to inhibit integrins and proteins of extracellular matrix binding.
Sequence:RGES
MW:447.5 Da
% peak area by HPLC:95
Storage condition:-20° C