Order Entry
Puerto Rico
ContactUsLinkComponent
Human alpha1(I) Collagen (614-639) Peptide
Human alpha1(I) Collagen (614-639) Peptide
Catalog # 103007-766
Supplier:  Anaspec Inc
CAS Number:  
Human alpha1(I) Collagen (614-639) Peptide
Catalog # 103007-766
Supplier:  Anaspec Inc
Supplier Number:  AS-63837
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    α1(I) Collagen Peptide|Type I Collagen Peptide|Collagen fibril inhibitor Peptide|α1(I) C-peptide
  • Protein/Peptide Name:
    alpha1(I) Collagen (614-639) Peptide
  • Purity:
    95%
  • Molecular Weight:
    2942.2 Da
  • Sequence:
    SAGFDFSFLPQPPQEKAHDGGRYYRA
  • Cat. No.:
    103007-766
  • Supplier no.:
    AS-63837

Specifications

About this item

This is a peptide inhibitor of collagen fibrillar matrix assembly.
Sequence:SAGFDFSFLPQPPQEKAHDGGRYYRA
MW:2942.2 Da
% peak area by HPLC:95
Storage condition:-20° C