Order Entry
Puerto Rico
ContactUsLinkComponent
FN-A208 Fusion Peptide
FN-A208 Fusion Peptide
Catalog # 103008-646
Supplier:  Anaspec
CAS Number:  
FN-A208 Fusion Peptide
Catalog # 103008-646
Supplier:  Anaspec
Supplier Number:  AS-65007
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Fibronectin-laminin fusion Peptide
  • Protein/Peptide Name:
    FN-A208 Fusion Peptide
  • Purity:
    95%
  • Molecular Weight:
    1716.9 Da
  • Sequence:
    GRGDSGAASIKVAVSADR
  • Cat. No.:
    103008-646
  • Supplier no.:
    AS-65007

Specifications

About this item

This peptide is a fusion of A208, derived from murine laminin a1, and the active site of fibronectin (GRGDS), with a glycine spacer. This peptide forms amyloid-like fibrils and promotes formation of actin stress fibers that mediate fibroblast cell attachment, offering it potential as a bioadhesive for tissue regeneration and engineering. FN-A208 interacts with IKVAV receptors and integrins. Its activity is disrupted by the presence of EDTA.
Sequence:GRGDSGAASIKVAVSADR
MW:1716.9 Da
% peak area by HPLC:95
Storage condition:-20° C