Order Entry
Puerto Rico
ContactUsLinkComponent
NFF-3, Mca-DNP
NFF-3, Mca-DNP
Catalog # 102996-800
Supplier:  Anaspec Inc
CAS Number:  
NFF-3, Mca-DNP
Catalog # 102996-800
Supplier:  Anaspec Inc
Supplier Number:  AS-27114
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Mca-DNP
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    NFF-3 MMP substrate|Substrate for MMP-3
  • Protein/Peptide Name:
    NFF-3
  • Purity:
    95%
  • Molecular Weight:
    1675.9 Da
  • Sequence:
    Mca-RPKPVE-Nva-WR-K(Dnp)-NH2
  • Cat. No.:
    102996-800
  • Supplier no.:
    AS-27114

Specifications

About this item

Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
NFF-3 is hydrolyzed rapidly by MMP-3 (kcat/Km = 218,000 s-1 M-1) and very slowly by MMP-9 (kcat/Km = 10,100 s-1 M-1), but no significant hydrolysis by MMP-1 and MMP-2. It is one of the few synthetic substrates selectively hydrolyzed by certain members of the MMP family and thus has important applications for the differentiation of MMP-3 activity from that of other MMPs. Abs/Em = 325/393 nm.
Sequence:Mca-RPKPVE-Nva-WR-K(Dnp)-NH2
MW:1675.9 Da
% peak area by HPLC:95
Storage condition:-20° C