Order Entry
Puerto Rico
ContactUsLinkComponent
 
520 MMP FRET Substrate XIV, QXL™ 520-FAM, AnaSpec
Catalog # 103005-942
Supplier:  Anaspec
CAS Number:  
520 MMP FRET Substrate XIV, QXL™ 520-FAM, AnaSpec
Catalog # 103005-942
Supplier:  Anaspec
Supplier Number:  AS-60581-01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    QXL™ 520-FAM
  • Size:
    0.1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    MMP Substrate XIV|MMP FRET|Matrix Metalloproteinase substrate XIV
  • Protein/Peptide Name:
    520 MMP FRET Substrate XIV
  • Purity:
    95%
  • Molecular Weight:
    1913.1 Da
  • Sequence:
    QXL™ 520 -γ-Abu-P-Cha-Abu-Smc-HA-Dab(5-FAM)-AK-NH2 (Smc=S-Methyl-L-cysteine)
  • Cat. No.:
    103005-942
  • Supplier no.:
    AS-60581-01

Specifications

About this item

Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
This peptide is a sensitive substrate for assaying MMP-1, 2, 3, 7, 8, 9, 12, and 13 activities, Abs/Em = 494/521 nm.
Sequence:QXL™ 520 -γ-Abu-P-Cha-Abu-Smc-HA-Dab(5-FAM)-AK-NH2 (Smc=S-Methyl-L-cysteine)
MW:1913.1 Da
% peak area by HPLC:95
Storage condition:-20° C