Order Entry
Puerto Rico
ContactUsLinkComponent
390 MMP FRET Substrate IV, Mca-DNP
390 MMP FRET Substrate IV, Mca-DNP
Catalog # 102996-792
Supplier:  Anaspec
CAS Number:  
390 MMP FRET Substrate IV, Mca-DNP
Catalog # 102996-792
Supplier:  Anaspec
Supplier Number:  AS-27109
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Mca-DNP
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    MMP FRET|MMP Substrate IV|Matrix Metalloproteinase substrate IV
  • Protein/Peptide Name:
    390 MMP FRET Substrate IV
  • Purity:
    95%
  • Molecular Weight:
    1100.2 Da
  • Sequence:
    Mca-P-Cha-G-Nva-HA-Dap(Dnp)-NH2
  • Cat. No.:
    102996-792
  • Supplier no.:
    AS-27109

Specifications

About this item

Matrix Metalloproteinases (MMPs) are a large family of endopeptidases. Collectively, MMPs can degrade all kinds of extracellular matrix proteins, and can also process a number of bioactive molecules. They are known to be involved in the cleavage of cell surface receptors, the release of apoptotic ligands, and chemokine/cytokine inactivation. MMPs are also thought to play a major role in cell behaviors such as cell proliferation, migration (adhesion/dispersion), differentiation, angiogenesis, apoptosis, and host defense.
This substrate is readily hydrolyzed by MMP-8, 9 and collagenase-3 (MMP-13), Abs/Em = 325/393 nm. The hydrolysis is inhibited in a 1:1 stoichiometric fashion by the tissue inhibitors of metalloproteinases, TIMP-1, TIMP-2, and TIMP-3.
Sequence:Mca-P-Cha-G-Nva-HA-Dap(Dnp)-NH2
MW:1100.2 Da
% peak area by HPLC:95
Storage condition:-20° C