Order Entry
Puerto Rico
ContactUsLinkComponent
Plants Phytochelatin 6
Plants Phytochelatin 6
Catalog # 103006-400
Supplier:  Anaspec Inc
CAS Number:  
Plants Phytochelatin 6
Catalog # 103006-400
Supplier:  Anaspec Inc
Supplier Number:  AS-61191
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Plants
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Gutathione-derived heavy metal-detoxifying Peptide|phytochelatin Peptide
  • Protein/Peptide Name:
    Phytochelatin 6
  • Purity:
    95%
  • Molecular Weight:
    1468.6 Da
  • Sequence:
    (γE-C)6-G
  • Cat. No.:
    103006-400
  • Supplier no.:
    AS-61191

Specifications

About this item

A glutathione-derived heavy metal-detoxifying peptide of higher plants consisting of 6 units of gammaGlu-Cys.
Sequence:(γE-C)6-G
MW:1468.6 Da
% peak area by HPLC:95
Storage condition:-20° C