Order Entry
Puerto Rico
ContactUsLinkComponent
[Succinyl-Asp6, NMePhe8]-Substance P (6-11)
[Succinyl-Asp6, NMePhe8]-Substance P (6-11)
Catalog # 102996-324
Supplier:  Anaspec Inc
CAS Number:  
[Succinyl-Asp6, NMePhe8]-Substance P (6-11)
Catalog # 102996-324
Supplier:  Anaspec Inc
Supplier Number:  AS-22887
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Substance P|NK3 tachykinin receptor agonist Peptide|Senktide
  • Protein/Peptide Name:
    [Succinyl-Asp6, NMePhe8]-Substance P (6-11)
  • Purity:
    95%
  • Molecular Weight:
    842.1 Da
  • Sequence:
    Suc-DF-(NMeF)-GLM-NH2
  • Cat. No.:
    102996-324
  • Supplier no.:
    AS-22887

Specifications

About this item

Senktide is a NK3 tachykinin receptor agonist. It was shown to induce bronchoconstriction in guinea pig lungs.
Sequence:Suc-DF-(NMeF)-GLM-NH2
MW:842.1 Da
% peak area by HPLC:95
Storage condition:-20° C