Substance P
About this item
Substance P (SP) is an important neuropeptide belonging to the tachykinin family. It acts as a neurotransmitter and neuromodulator. It is closely related to neurokinin A, both originating from the same precursor preprotachykinin A. It is released from the terminals of sensory nerves and is involved in inflammation and pain processes.
Sequence:RPKPQQFFGLM-NH2
MW:1347.7 Da
% peak area by HPLC:95
Storage condition:-20° C
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Substance P (SP) is an important neuropeptide belonging to the tachykinin family. It acts as a neurotransmitter and neuromodulator. It is closely related to neurokinin A, both originating from the same precursor preprotachykinin A. It is released from the terminals of sensory nerves and is involved in inflammation and pain processes.
Sequence:RPKPQQFFGLM-NH2
MW:1347.7 Da
% peak area by HPLC:95
Storage condition:-20° C
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 102996-504
- 102996-506
- Protein/Peptide Name:
- Substance P
- Substance P
- Protein Synonyms:
- Substance P neuropeptide
- Substance P neuropeptide
- Conjugation:
- Unconjugated
- Unconjugated
- Sequence:
- RPKPQQFFGLM-NH2
- RPKPQQFFGLM-NH2
- Purity:
- 95%
- 95%
- Molecular Weight:
- 1347.7 Da
- 1347.7 Da
- Storage Conditions:
- –20 °C
- –20 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: AS-24279Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102996-504Protein/Peptide NameSubstance PProtein SynonymsSubstance P neuropeptideConjugationUnconjugatedSequenceRPKPQQFFGLM-NH2Purity95%Molecular Weight1347.7 DaStorage Conditions–20 °CMarketsource - Supplier Number: AS-24280Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102996-506Protein/Peptide NameSubstance PProtein SynonymsSubstance P neuropeptideConjugationUnconjugatedSequenceRPKPQQFFGLM-NH2Purity95%Molecular Weight1347.7 DaStorage Conditions–20 °CMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



