Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Size:25 mg
- Storage Conditions:–20 °C
- Protein Synonyms:Leu- Enkephalin Peptide|Enkephalin Peptide
- Protein/Peptide Name:[Leu5]-Enkephalin
- Purity:95%
- Molecular Weight:555.7 Da
- Sequence:YGGFL
- Cat. No.:102996-548
- Supplier no.:AS-24333
Specifications
About this item
Enkephalins are pentapeptides involved in regulating nociception in the body. There are two enkephalin forms, one containing leucine and the other containing methionine ("met"). Both are products of the proenkephalin gene.
Sequence:YGGFL
MW:555.7 Da
% peak area by HPLC:95
Storage condition:-20° C