Order Entry
United States
ContactUsLinkComponent
 
5-Fluoroorotic acid monohydrate (5-FOA) 100 mg/mL in DMSO , 98%, liquid
Catalog # 77001-956
Supplier:  Zymo Research
CAS Number:  
5-Fluoroorotic acid monohydrate (5-FOA) 100 mg/mL in DMSO , 98%, liquid
Catalog # 77001-956
Supplier:  Zymo Research
Supplier Number:  F9003
CAS Number:  

Specifications

  • Size:
    10 ml
  • Environmentally Preferable:
  • Cat. No.:
    77001-956

Specifications

About this item

Synonyms: 5-FOA, 5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-Fluorouracil-6-carboxylic acid monohydrate
 :
 :174.0 g/mol (anhy)
 :GHS07
Storage Temperature:Freezer
CAS Number:220141-70-8
 :703-95-7
Formula:C₅H₃FN₂O₄·1H₂O
MDL Number:MFCD00150658