ContactUsLinkComponent
Static-Safe Wipe, Clean Room Grade, Large, MicroCare
Static-Safe Wipe, Clean Room Grade, Large, MicroCare
Catalog # 76020-250
Supplier:  MicroCare
undefined
Static-Safe Wipe, Clean Room Grade, Large, MicroCare
Catalog # 76020-250
Supplier:  MicroCare
Supplier Number:  MCC-W99CY
Specifications
  • Description:
    Static-Safe Wipe, Clean Room Grade, Large
  • Cat. No.:
    76020-250
Specifications
About this item
This is an excellent quality, large knitted wipe.
  • ESD-safe wipe
  • Embedded conductive fibers
It is unique because it has carbon fibers knitted into the fabric so the wipe can dissipate static even when used dry. The fabric is heavier than the standard W99 wipe, so it picks up more contamination faster, speeding cleaning. It is pre-washed, so it cleans better and lints less. The edges are heat-sealed which also makes the wipe a better cleaning choice. Lastly, the wipes are packaged in a clean room, in ESD-safe bags, and is double-bagged for Class 10 clean rooms.
Packaging: Class 10 Clean Room Packaged; 150 sheets/bag, 5 bags/case.