ContactUsLinkComponent
SMT Stencil Wipe, Electronics Grade, Large, MicroCare
SMT Stencil Wipe, Electronics Grade, Large, MicroCare
Catalog # 76020-249
Supplier:  MicroCare
undefined
SMT Stencil Wipe, Electronics Grade, Large, MicroCare
Catalog # 76020-249
Supplier:  MicroCare
Supplier Number:  MCC-W99
Specifications
  • Description:
    SMT Stencil Wipe, Electronics Grade, Large
  • Dimensions:
    23 x 23 cm
  • Cat. No.:
    76020-249
Specifications
About this item
This is an excellent, medium quality wipe sized for cleaning SMT stencils and other larger chores.
  • SMT stencil wipe, medium quality, medium price, large size
  • Smooth, white, medium absorbency
  • Relatively lint-free
  • Good purity
  • Economical polyester-and-cellulose fabric
  • Optimal size for cleaning large devices, like SMT stencil cleaning
Like it’s smaller sibling, the W66 wipe, the W99 is a perennial MicroCare best-seller. Polyester and cellulose fabric. Medium absorption, medium weight, relatively lint-free, good purity. It’s the optimal size for use on stencils and other maintenance chores like cleaning pallets and reflow ovens, but it’s probably too big to use on most modern PCBs.
Packaging: 300 sheets/bag, 12 bags/case.