Order Entry
Puerto Rico
ContactUsLinkComponent
SMT Stencil Wipe, Economy Grade, Large, MicroCare
SMT Stencil Wipe, Economy Grade, Large, MicroCare
Catalog # 76020-235
Supplier:  MicroCare
undefined
SMT Stencil Wipe, Economy Grade, Large, MicroCare
Catalog # 76020-235
Supplier:  MicroCare
Supplier Number:  MCC-W11

Specifications

  • Color:
    White
  • Description:
    SMT Stencil Wipe, Economy Grade, Large
  • Dimensions:
    21 x 28 cm
  • Cat. No.:
    76020-235
  • Packaging:
    100 Sheets/Bag

Specifications

About this item

This is a good quality stencil wipe that offers excellent cleaning at an economical price.

  • SMT stencil wipe, economy quality with large size
  • Smooth, white, medium absorbency
  • Optimal size for cleaning large devices, like SMT stencil cleaning

It’s a large with, about 8 inches by 11", so it delivers lots of scrubbing. It is made from hydroentangled polyester and cellulose, without any glues or binders for better cleaning. The material is lighter than other choices, so it’s a better choice for less heavily contaminated surfaces. This wipe is packaged with the MicroCare reflow oven cleaner (#MCC-ROC) and the MicroCare aqueous stencil cleaner (#MCC-BGA).

Packaging: 100 sheets/bag, 6 bags/case.