Orders LinkContactUsLinkComponent
Gold Sol, Electron Microscopy Sciences
Gold Sol, Electron Microscopy Sciences
Marketsource
Gold Sol, Electron Microscopy Sciences
Gold Sol, Electron Microscopy Sciences
Solutions of high quality unconjugated gold particles.
  • Particle sizes of 0.8, 6, 10 ,15 and 25 nm
  • Guaranteed shelf life of 12 months
  • Monodisperse particle population
To prepare a high quality (immuno) gold conjugate it is important to have particles with uniform size and highly adsorptive surfaces.
Introduction
The preparation of conventional gold reagents is based on gold particles with a diameter suited for direct electron microscopic visualization. Aurion offers a range of these Reagents which cover most of the approaches in transmission and scanning electron microscopy.
Aurion Gold Sols (solutions of high quality unconjugated gold particles) provide opportunities for users to prepare conjugates with primary antibodies, ligands and other binding agents with the same particle characteristics as in the Conventional Immunogold Reagents.
Product Description
AURION Gold Sols are prepared according to a unique protocol, warranting narrow size distribution and reproducible adsorption characteristics.
The AURION gold sols are available in the same size range as the Conventional Immunogold Reagents: 6, 10, 15 and 25 nm. The particle population is monodispersed and thus shows minimal size variation and overlap. Typically, the coefficient of variance for the 6 and 25 nm particle size sols is less than 12%, whereas the 10 and 15 nm size sols show less than 10% variation. Actual lot specifications (size, variation and expiry date) are indicated on the accompanying package insert.
Package size: 100 ml of high quality gold sol at an OD520 nm of approximately 1.
Storage
AURION gold sols have a guaranteed shelf life of 12 months from the date of quality control analysis.
The products should be stored at 4 – 8°C.
Freezing is not recommended.
  • Catalog No:
  • 100487-924
  • 100487-926
  • 100487-928
  • 100487-930
  • 102091-022
  • Antibody Type:
  • Secondary
  • Secondary
  • Secondary
  • Secondary
  • Secondary
Product Family Options
  • Description
    Volume
    Size
  • Gold Sol
    100 ml
    UltraSmall
    Each (100ml)
    Each (100ml) - $443.48
    $443.48
    Each (100ml)
       $443.48
     : 25510
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    100487-924
    Antibody Type
    Secondary
    Minority or Woman-Owned Business Enterprise
  • Gold Sol
    100 ml
    6 nm
    Each (100ml)
    Each (100ml) - $443.48
    $443.48
    Each (100ml)
       $443.48
     : 25512
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    100487-926
    Antibody Type
    Secondary
    Minority or Woman-Owned Business Enterprise
  • Gold Sol
    100 ml
    10 nm
    Each (100ml)
    Each (100ml) - $443.48
    $443.48
    Each (100ml)
       $443.48
     : 25514
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    100487-928
    Antibody Type
    Secondary
    Minority or Woman-Owned Business Enterprise
  • Gold Sol
    100 ml
    15 nm
    Each (100ml)
    Each (100ml) - $443.48
    $443.48
    Each (100ml)
       $443.48
     : 25516
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    100487-930
    Antibody Type
    Secondary
    Minority or Woman-Owned Business Enterprise
  • Gold Sol
    100 ml
    25 nm
    Each
    Each - $451.36
    $451.36
    Each
       $451.36
     : 25509
    Marketsource
    Specifications
    Cat. No.
    102091-022
    Antibody Type
    Secondary
    Marketsource
This product has been blocked by your organization. Please contact your purchasing department for more information