Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences

Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences

Thin Specimen Split Mount, 1/2"
Catalog # 103302-548
Supplier #:  75950-08
Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences
Catalog # 103302-548
Thin Specimen Split Mount on Pin Stub, Electron Microscopy Sciences

Thin Specimen Split Mount, 1/2"

Price
$63.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Thin specimen split mount on pin stub is used for cross section examination in scanning electron microscopes.

  • Used for paper, metals, plastic, textiles, plant material and more
  • Machined aluminum with stainless steel allen set screws
  • Opens to 6.4 mm
  • Grooved head

Height of head is 7.4 mm (0.29"), Pin Length is 8 mm (0.314") and pin is centered.

Delivery information: Includes allen wrench.

Specifications
  • Description:
    Thin Specimen Split Mount
  • Size:
    1/2"

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.