To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:TAMRA (5-Carboxytetramethylrhodamin)
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 °C
- Protein Synonyms:α-melanocyte-stimulating hormone analog Peptide|NDP-MSH Peptide|α-MSH analog Peptide|α-MSH TAMRA|alpha-Melanotropin analog Peptide
- Protein/Peptide Name:NDP-MSH, 5-TAMRA-labeled
- Purity:95%
- Molecular Weight:2017.3 Da
- Sequence:5-TMR-SYS-Nle-EHfRWGKPV-NH2
- Cat. No.:103003-236
- Supplier no.:AS-60564-1
Specifications
About this item
This NDP-MSH peptide is fluorescently (5-TAMRA)-labeled on the N-terminus. Abs/Em=544/572 nm. NDP-MSH is a synthetic analog of alpha-Melanotropin (alpha-MSH, alpha-melanocyte-stimulating hormone) in which Asp4 has been replaced with Norleucine and L-Phe with the D-isomer of Phe [Nle4D-Phe7] (NDP). NDP-MSH is a more potent in increasing pigmentation and decreasing UV damage in fair-skinned Caucasian volunteers.
Sequence: 5-TMR-SYS-Nle-EHfRWGKPV-NH2
MW: 2017.3 Da
% Peak area by HPLC: 95
Storage condition: -20° C