Order Entry
Puerto Rico
ContactUsLinkComponent
Alpha9-Gliadin (57-68)
Alpha9-Gliadin (57-68)
Catalog # 103008-718
Supplier:  Anaspec Inc
CAS Number:  
Alpha9-Gliadin (57-68)
Catalog # 103008-718
Supplier:  Anaspec Inc
Supplier Number:  AS-65105
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    α9 Gliadin|Gliadin Peptide|Gliadin 57-68
  • Protein/Peptide Name:
    alpha9-Gliadin (57-68)
  • Purity:
    95%
  • Molecular Weight:
    1455.7 Da
  • Sequence:
    QLQPFPQPQLPY
  • Cat. No.:
    103008-718
  • Supplier no.:
    AS-65105

Specifications

About this item

Gliadin is a gluten component. This peptide is derived from amino acid residues 57-68 of a9-gliadin and represents an immunodominant epitope that has been shown to elicit HLA-DQ2-restricted T-cell responses in celiac patients. This epitope is resistant to pancreatic proteolysis.
Sequence: QLQPFPQPQLPY
MW: 1455.7 Da
% Peak area by HPLC: 95
Storage condition: -20° C