Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Size:1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:EPO-mimetic FITC|EPO-mimetic Peptide FITC|Erythropoeitin mimetic Peptide FITC labeled
- Protein/Peptide Name:EMP17, FITC-LC labeled
- Purity:95%
- Molecular Weight:2483.9 Da
- Sequence:FITC-LC-TYSCHFGPLTWVCKPQGG
- Cat. No.:103002-744
- Supplier no.:AS-60180-1
This is a fluorescent (FITC)-labeled Erythropoietin (EPO)-mimetic peptide (EMP17), Abs/Em=494/520 nm. Erythropoietin (EPO) is the hormone involved in red blood cell production, which activates its receptor by binding to the receptor's extracellular domain and presumably dimerizing two receptor monomers to initiate signal transduction. EMP contains two potentially reactive amines, one at the amino terminus of the peptide and one in the side chain of the single lysine within the peptide sequence.
Sequence: FITC-LC-TYSCHFGPLTWVCKPQGG
MW: 2483.9 Da
% Peak area by HPLC: 95
Storage condition: -20° C