Order Entry
Puerto Rico
ContactUsLinkComponent
Gap 27
Gap 27
Catalog # 103008-436
Supplier:  Anaspec Inc
CAS Number:  
Gap 27
Catalog # 103008-436
Supplier:  Anaspec Inc
Supplier Number:  AS-64765
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein/Peptide Name:
    Gap 27
  • Purity:
    95%
  • Molecular Weight:
    1304.6 Da
  • Sequence:
    REKIITSFIPT
  • Cat. No.:
    103008-436
  • Supplier no.:
    AS-64765

Specifications

About this item

This peptide is a scrambled version of the Gap 27 domain of Connexin.
Sequence:REKIITSFIPT
MW:1304.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C