Order Entry
Puerto Rico
ContactUsLinkComponent
[Pyr1]-Apelin-13
[Pyr1]-Apelin-13
Catalog # 103005-982
Supplier:  Anaspec Inc
CAS Number:  
[Pyr1]-Apelin-13
Catalog # 103005-982
Supplier:  Anaspec Inc
Supplier Number:  AS-60833
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Pyroglutamate Apelin 13|Apelin 13 pyroglutamate|Pyr1-Apelin 13
  • Protein/Peptide Name:
    [Pyr1]-Apelin-13
  • Purity:
    95%
  • Molecular Weight:
    1533.8 Da
  • Sequence:
    Pyr-RPRLSHKGPMPF-OH
  • Cat. No.:
    103005-982
  • Supplier no.:
    AS-60833

Specifications

About this item

[125I]-(Pyr1)Apelin-13 binds with high affinity and selectivity to human cardiac tissue.
Sequence:Pyr-RPRLSHKGPMPF-OH
MW:1533.8 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C