Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Atrial Natriuretic Peptide (4-18)
Rat Atrial Natriuretic Peptide (4-18)
Catalog # 103006-666
Supplier:  Anaspec
CAS Number:  
Rat Atrial Natriuretic Peptide (4-18)
Catalog # 103006-666
Supplier:  Anaspec
Supplier Number:  AS-62839
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Rat
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    ANP 4-18|ANP 4-18 rat
  • Protein/Peptide Name:
    Atrial Natriuretic Peptide (4-18)
  • Purity:
    95%
  • Molecular Weight:
    1594.9 Da
  • Sequence:
    RSSCFGGRIDRIGAC-NH2 (Disulfide bridge 4-15)
  • Cat. No.:
    103006-666
  • Supplier no.:
    AS-62839

Specifications

About this item

This peptide corresponds to amino acids 4 to 18 of rat atrial natriuretic peptide (ANP). It has been used as a cystein containing peptide model in quantitative mass spectroscopic analysis. This fragment is also related to C-ANP (4-23);  (Des-Gln18,des-Ser19,des-Gly20,22,des-Leu21), which is completely selective in discriminating rat C-AMP receptors.
Sequence:RSSCFGGRIDRIGAC-NH2 (Disulfide bridge 4-15)
MW:1594.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C