About this item
Rat ANP differs from the human hormone by only one residue at position 12. ANP (1-28) peptide hormone constitutes the first 28 amino acids of the ANP synthesized in the heart of different vertebrates. It plays an important role in the regulation of blood pressure and natriuresis/diuresis. Residues Phe8, Arg14 and the C-terminal sequence of ANP are known to bind to human NPR-A (Natriuretic peptide receptor type A).
Sequence:SLRRSSCFGGRIDRIGAQSGLGCNSFRY (Disulfide bridge: 7-23)
MW:3062.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Species:Rat
- Storage Conditions:–20 °C
- Protein Synonyms:ANP 1-28|ANP rat
- Protein/Peptide Name:Atrial Natriuretic Peptide (1-28)
- Purity:95%
- Molecular Weight:3062.5 Da
- Sequence:SLRRSSCFGGRIDRIGAQSGLGCNSFRY (Disulfide bridge: 7-23)