Order Entry
Puerto Rico
ContactUsLinkComponent
Plasmin Substrate, AFC (Antibody-fluorophore conjugate)
Plasmin Substrate, AFC (Antibody-fluorophore conjugate)
Catalog # 102996-444
Supplier:  Anaspec Inc
CAS Number:  
Plasmin Substrate, AFC (Antibody-fluorophore conjugate)
Catalog # 102996-444
Supplier:  Anaspec Inc
Supplier Number:  AS-24133
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    AFC (Antibody-fluorophore conjugate)
  • Size:
    10 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Plasmin substrate|fluorescent plasmin substrate|Plasmin AFC substrate
  • Protein/Peptide Name:
    Plasmin Substrate
  • Purity:
    95%
  • Molecular Weight:
    569.5 Da
  • Sequence:
    vLK-AFC
  • Cat. No.:
    102996-444
  • Supplier no.:
    AS-24133

Specifications

About this item

This is a fluorescent plasmin substrate, Abs/Em=380/500 nm.
Sequence:vLK-AFC
MW:569.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C