Order Entry
Puerto Rico
ContactUsLinkComponent
Thrombin Substrate
Thrombin Substrate
Catalog # 103007-720
Supplier:  Anaspec Inc
CAS Number:  
Thrombin Substrate
Catalog # 103007-720
Supplier:  Anaspec Inc
Supplier Number:  AS-63776
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    Chrmogenic thrombin substrate|Thrombin substrate|Thrombin S2238 substrate|Chromogenic substrate
  • Protein/Peptide Name:
    Thrombin Substrate
  • Purity:
    95%
  • Molecular Weight:
    552.6 Da
  • Sequence:
    f - Pip - R - pNA
  • Cat. No.:
    103007-720
  • Supplier no.:
    AS-63776

Specifications

About this item

This is a chromogenic substrate for thrombin, Abs=405 nm.
Sequence:f - Pip - R - pNA
MW:552.6 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C