Order Entry
Puerto Rico
ContactUsLinkComponent
Chicken OVA (323-339)
Chicken OVA (323-339)
Catalog # 103007-398
Supplier:  Anaspec Inc
CAS Number:  
Chicken OVA (323-339)
Catalog # 103007-398
Supplier:  Anaspec Inc
Supplier Number:  AS-62571-5
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Chicken
  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    OVA 323-339 Peptide|OVA Peptide|OVA 323-339 amide
  • Protein/Peptide Name:
    OVA (323-339)
  • Purity:
    95%
  • Molecular Weight:
    1773 Da
  • Sequence:
    ISQAVHAAHAEINEAGR-NH2
  • Cat. No.:
    103007-398
  • Supplier no.:
    AS-62571-5

Specifications

About this item

This peptide is amino acids 323 to 339 amidated fragment of ovalbumin (OVA), the H-2b-restricted OVA class II epitope. This peptide is recognized by many T cells because it contains multiple T cell epitopes. This OVA fragment contains a nested set of CD4+ T cell epitopes. OVA 323 to 339 can be presented by I-Ad in at least three binding registers. The residues 327 to 333 are critical for peptide binding to I-Ad.
Sequence: ISQAVHAAHAEINEAGR-NH2
MW: 1773 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C