Order Entry
Puerto Rico
ContactUsLinkComponent
Human LL-37 fragment (18-37)
Human LL-37 fragment (18-37)
Catalog # 103007-518
Supplier:  Anaspec Inc
CAS Number:  
Human LL-37 fragment (18-37)
Catalog # 103007-518
Supplier:  Anaspec Inc
Supplier Number:  AS-63712
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    –20 °C
  • Protein Synonyms:
    LL-37 fragment|LL-37 (18-37)|LL 18-37
  • Protein/Peptide Name:
    LL-37 fragment (18-37)
  • Purity:
    95%
  • Molecular Weight:
    2468.9 Da
  • Sequence:
    KRIVQRIKDFLRNLVPRTES
  • Cat. No.:
    103007-518
  • Supplier no.:
    AS-63712

Specifications

About this item

This is fragment 18-37 of LL-37, it exhibits enhanced antimicrobial activity. Antimicrobial peptide LL-37, belonging to the cathelicidin family, is the first amphipathic alpha-helical peptide isolated from human. It plays an important role in the first line of defense (innate immunity) against local infection and systemic invasion of pathogens at sites of inflammation and wounds.
Sequence: KRIVQRIKDFLRNLVPRTES
MW: 2468.9 Da
% Peak Area by HPLC: ≥ 95%
Storage condition: -20°C