ContactUsLinkComponent
3M™ OCS System for Vending Applications featuring VH3 Valve-in-Head Design, Model VH3/VEN351-K, 5626209
3M™ OCS System for Vending Applications featuring VH3 Valve-in-Head Design, Model VH3/VEN351-K, 5626209
Catalog # 76030-527
Supplier:  3M Healthcare
3M™ OCS System for Vending Applications featuring VH3 Valve-in-Head Design, Model VH3/VEN351-K, 5626209
Catalog # 76030-527
Supplier:  3M Healthcare
Supplier Number:  5626209
Specifications
  • Cat. No.:
    76030-527
Specifications
About this item
Sanitary quick change system with built-in valve-in-head. For general use.
  • No external shut-off valve required
  • 3/8" quick connect fittings
  • 1 micron sediment reduction
  • Premium performance carbon block does not require activation
  • Chlorine taste & odor reduction
  • Built-in scale inhibitor and includes KDF® which is a bacteriostatic media