Order Entry
Puerto Rico
ContactUsLinkComponent
3M™ Zeta Plus™ 47 mm Filter Housing
3M™ Zeta Plus™ 47 mm Filter Housing
Catalog # 76031-922
Supplier:  3M Healthcare
3M™ Zeta Plus™ 47 mm Filter Housing
Catalog # 76031-922
Supplier:  3M Healthcare
Supplier Number:  7000001638

Specifications

  • Material:
    Stainless steel 316L
  • Cat. No.:
    76031-922
  • Model:
    7022501A
  • Inlet size:
    Mini sanitary

Specifications

About this item