3M™ Zeta Plus™ C Series Filter Cartridge
Catalog # 76028-925
Supplier: 3M Healthcare
Specifications
- Cat. No.:76028-925
- Model:4524401B10C
Specifications
About this item
We’ve designed Zeta Plus™ C Series Cartridges to provide optimal clarification of food and beverage fluids.
- Depth filtration media designed to retain contaminants by mechanical entrapment and electrokinetic adsorption
- Positively charged filter uses adsorption for additional particle containment
Our Zeta Plus™ C Series Sheets are designed to retain contaminants by way of mechanical entrapment and electrokinetic adsorption. The positively charged filter uses absorption for additional particle containment.