ContactUsLinkComponent
3M™ Commercial Reverse Osmosis Membrane for Model FSTM-075, 5598728
3M™ Commercial Reverse Osmosis Membrane for Model FSTM-075, 5598728
Catalog # 76030-251
Supplier:  3M Healthcare
3M™ Commercial Reverse Osmosis Membrane for Model FSTM-075, 5598728
Catalog # 76030-251
Supplier:  3M Healthcare
Supplier Number:  7100068022
Specifications
  • Cat. No.:
    76030-251
  • Model:
    5598728
Specifications
About this item
The 3M™ Commercial Reverse Osmosis Membrane for Model FSTM-075 has a Sanitary Quick Change (SQC) design allows for easy cartridge change-outs.
  • Provides consistent quality water
  • Reverse Osmosis replacement membrane for TDS reduction in boilerless steamers, combi-ovens and flash steamers
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench
  • Replacement RO Membrane for Model FSTM-075 Reverse Osmosis Systems
3M™ ScaleGard™ Reverse Osmosis Replacement Filter Cartridge, Model CFS M is designed to reduce total dissolved solids (TDS) in boilerless steamers, combi-ovens and flash steamers. Our Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench.