ContactUsLinkComponent
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112, 5631605
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112, 5631605
Catalog # 76030-579
Supplier:  3M Healthcare
undefined
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112, 5631605
Catalog # 76030-579
Supplier:  3M Healthcare
Supplier Number:  7100044900
Specifications
  • Cat. No.:
    76030-579
  • Model:
    5631605
Specifications
About this item
3M™ Water Filtration Products Model CFS9112 Retrofit 9000 Series Replacement Cartridge provides Recipe Quality Water™ for consistent cold beverages by reducing undesirable sediment,* and chlorine taste and odor. Our retrofit replacement fits most Everpure® commercial foodservice heads allowing you to choose from a variety of trusted 3M™ Water Filtration Products commercial 9000 series cartridges.
  • Provides consistent quality water for commercial post mix machines by reducing sediment,* and chlorine taste and odor
  • Enhances water clarity for visually appealing cold beverages
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products Model CFS9112 Retrofit 9000 Series Replacement Cartridge is designed to be effective at reducing sediment*, and chlorine taste and odor, allowing you to feel more confident in the quality of the products you serve