ContactUsLinkComponent
3M™ High Flow Series Replacement Cartridge, Model HF37, 5632302
Catalog # 76030-621
Supplier:  3M Healthcare
3M™ High Flow Series Replacement Cartridge, Model HF37, 5632302
Catalog # 76030-621
Supplier:  3M Healthcare
Supplier Number:  7100049863
Specifications
  • Cat. No.:
    76030-621
  • Model:
    5632302
Specifications
About this item
3M™ high flow series replacement cartridge, model HF37 provides consistent quality water by reducing particulate* and chlorine taste and odor from incoming water. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench.
  • Provides consistent quality water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench