ContactUsLinkComponent
3M™ Replacement Cartridge OCS Retrofit, Model CS-24, 5631510
3M™ Replacement Cartridge OCS Retrofit, Model CS-24, 5631510
Catalog # 76030-569
Supplier:  3M Healthcare
undefined
3M™ Replacement Cartridge OCS Retrofit, Model CS-24, 5631510
Catalog # 76030-569
Supplier:  3M Healthcare
Supplier Number:  7100049843
Specifications
  • Cat. No.:
    76030-569
  • Model:
    5631510
Specifications
About this item
3M™ Water Filtration Products Model CS-24 OCS Retrofit Replacement Cartridge provides consistent high quality water for beverages by reducing undesirable sediment*, chlorine taste and odor and cyst** from incoming water. Scale* inhibition helps to protect equipment from the potentially harmful effects of scale build-up. This cartridge is compatible with most OCS Everpure® foodservice heads.
  • Provides consistent high quality water for hot beverage applications
  • Reduces sediment*, chlorine taste and odor and cyst** from incoming water
  • Built-in scale* inhibition as an economical solution to address the potential of scale build-up in hot beverage applications
  • Compatible with most OCS Everpure® foodservice heads
Our 3M™ Water Filtration Products Model CS-24 OCS Retrofit Replacement Cartridge is designed to be effective at reducing sediment*, chlorine taste and odor, and cyst**, allowing you to feel more confident in the quality of your water.
Scale* inhibition helps to protect your equipment from the potentially harmful effects of scale build-up. This filter cartridge is compatible with most OCS Everpure® foodservice heads, and various 3M™ Water Filtration Products OCS retrofit cartridges are available to meet particular water quality concerns. This cartridge's high quality water filtration capabilities can make it a great option for your office hot beverage needs.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A History of Innovation.
*As tested and verified by manufacturer's laboratory.
**Based on the use of Cryptosporidium parvum oocysts.
Everpure is a trademark of PENTAIR FILTRATION SOLUTIONS, LLC