- Cat. No.:76030-567
- Model:5631509
3M™ Water Filtration Products Model CS-15 OCS Retrofit Replacement Cartridge provides consistent quality water for cold beverages by reducing sediment* and chlorine taste and odor from incoming water. This filter cartridge is compatible with most OCS Everpure® foodservice heads.
- Provides consistent quality water by reducing sediment* and chlorine taste and odor for drinking water applications
- Reduces sediment* and chlorine taste and odor from incoming water
- Compatible with most OCS Everpure® foodservice heads
Our 3M™ Water Filtration Products Model CS-15 OCS Retrofit Replacement Cartridge is designed to be effective at reducing sediment* and chlorine taste and odor, allowing you to feel more confident in the quality of your water.
This filter cartridge is compatible with most OCS Everpure® foodservice heads, and various 3M™ Water Filtration Products OCS retrofit cartridges are available to meet particular water quality concerns. These features together can make this replacement cartridge a great option for your office cold beverage needs.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A History of Innovation
*As tested and verified by manufacturer's laboratory.
Everpure is a trademark of PENTAIR FILTRATION SOLUTIONS, LLC