Specifications
- Cat. No.:76030-511
- Model:5626106
Specifications
About this item
3M™ water filtration products model HC351-S OCS office coffee replacement cartridge provides consistent quality water for hot beverages by reducing undesirable sediment* chlorine taste and odor and scale* from incoming water.
- Provides consistent high quality water for office coffee and hot cup applications by reducing fine sediment*, chlorine taste and odor, asbestos and cyst1
- Reduces the ability of calcium and magnesium to form hard scale* helping to reduce downtime and unscheduled service maintenance
- Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
- Replacement cartridge for VH3/HC350-S System
Our 3M™ water filtration products model HC351-S OCS office coffee replacement cartridge is designed to be effective at reducing sediment,* scale* and chlorine taste and odor allowing you to feel more confident in the quality of the products you serve.
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs without the need for a filter wrench and are sanitary by design, requiring no contact with the filter media during cartridge change-out. Replacement cartridge for use in VH3/HC351-S system.
3M™ water filtration products replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M water filtration products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
Recipe Quality Water™
A history of innovation
For more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine and other applications.
*Tested and verified by manufacturer's laboratory.
1As tested with E. coli ATCC (11229). Tested and verified by manufacturer's laboratory.