ContactUsLinkComponent
3M™ Replacement Cartridge OCS Retrofit, Model CS-25, 5631511
3M™ Replacement Cartridge OCS Retrofit, Model CS-25, 5631511
Catalog # 76030-571
Supplier:  3M Healthcare
3M™ Replacement Cartridge OCS Retrofit, Model CS-25, 5631511
Catalog # 76030-571
Supplier:  3M Healthcare
Supplier Number:  7100049844
Specifications
  • Cat. No.:
    76030-571
  • Model:
    5631511
Specifications
About this item
3M™ waer filtration products model CS-25 OCS retrofit replacement cartridge provides consistent high quality water for cold beverages by reducing sediment*, chlorine taste and odor and cyst** from incoming water. This filter cartridge is compatible with most OCS Everpure® foodservice heads.
  • Provides consistent quality water for cold beverage applications
  • Reduces sediment*, chlorine taste and odor and cyst** from incoming water
  • Compatible with most OCS Everpure® foodservice heads
Our 3M™ water filtration products model CS-25 OCS retrofit replacement cartridge is designed to be effective at reducing sediment*, chlorine taste and odor and cyst**, allowing you to feel more confident in the quality of your water.
This filter cartridge is compatible with most OCS Everpure® foodservice heads, and various 3M™ water filtration products OCS retrofit cartridges are available to meet particular water quality concerns. This cartridge's high quality water filtration capabilities can make it a great option for your office cold beverage needs.
3M™ water fltration products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A History of innovation
*As tested and verified by manufacturer's laboratory.
**Based on the use of Cryptosporidium parvum oocysts.
Everpure is a trademark of PENTAIR FILTRATION SOLUTIONS, LLC.