3M™ CUNO Legacy Drop-In Style Replacement Cartridge, Model CFS217, 5578606
Catalog # 76030-205
Supplier: 3M Healthcare
Specifications
- Cat. No.:76030-205
- Model:5578606
Specifications
About this item
3M™ CUNO legacy drop–in style replacement cartridge, model CFS217, which is used in model CFS200, provides consistent quality water by reducing chlorine taste and odor and particulate from incoming water.
- Drop–in style replacement cartridge
- Provides consistent quality water
- Reduces particulate and chlorine taste and odor from incoming water
- Designed with high–adsorption granular activated carbon
- For use in Model CFS200 filtration system
This space saving, compact design contains a high–adsorption granular activated carbon.