ContactUsLinkComponent
3M™ Scale Inhibition System, Model SF18-S, 6 per case, 5607708
3M™ Scale Inhibition System, Model SF18-S, 6 per case, 5607708
Catalog # 76030-279
Supplier:  3M Healthcare
3M™ Scale Inhibition System, Model SF18-S, 6 per case, 5607708
Catalog # 76030-279
Supplier:  3M Healthcare
Supplier Number:  7100051040
Specifications
  • Cat. No.:
    76030-279
  • Model:
    5607708
Specifications
About this item
3M™ Water Filtration Products High Flow Series (-S) model filter system provide Recipe Quality Water™ for commercial ice machines and commercial steam tables by reducing the effects of sediment*, chlorine taste and odor and scale* while helping you protect your equipment.
  • Provides consistent quality water for commercial ice machines and commercial steam tables by reducing sediment*, chlorine taste and odor and scale*
  • Enhances water clarity for visually appealing iced beverages
  • Protects equipment from sediment*, chlorine and scale*
  • Reduces chlorine and corrosion potential and helps extend equipment life
  • Reduces calcium and magnesium formation as hard scale* for reduced downtime and unscheduled maintenance
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy change-outs
  • Integrated Membrane Pre-Activated Carbon Technology (“I.M.P.A.C.T”) provides superior cartridge life for models HF90-S, HF60-S, HF60-CLS, and HF40-S
This High Flow Series (-S) line of filter cartridges are sanitary by design, requiring no physical contact with the filter media during cartridge change-out
Our Sanitary Quick Change (SQC) feature allows for fast and easy cartridge change-outs without the need for a filter wrench. Other smart design details include our integral mounting bracket with keyhole cut out design, which helps add stability and provides less stress on plumbing lines when mounted to a wall or adjacent piece of equipment. Some High Flow Series (-S) replacement cartridge filters are designed to be effective at reducing common water-borne bacteria** and cyst¹ for models HF90-S, HF90-S-SR, HF90-S-SR5, HF60-S, HF60-CLS, HF40-S, helping you feel more confident in the quality of the products you serve. Hard scale can accumulate and clog distribution nozzles and evaporator plates in commercial ice machines and coat heating tubes or elements in steam tables; increasing maintenance and downtime. The added benefit of our built-in scale inhibition helps reduce calcium and magnesium from precipitating on metal components as hard scale*.
Model HF-S Series Best Features
Filter cartridges are o-ring seal type
Cartridge body is constructed from glass-filled polypropylene
Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change-out
Filter cartridges require no pre-activation
In many filtration applications, carbon is often the preferred media of choice to absorb impurities. Carbon is highly effective due to the vast surface area contained within a small footprint. As effective as carbon is, activated carbon is even more effective. Activated carbon is made of small clusters of carbon atoms, and is produced by heating the carbon source (coal, lignite, wood, nutshells or peat) in the absence of air. Adsorption is the adhesion of atoms, ions, or molecules from a gas, liquid, or dissolved solid to a surface. This process differs from absorption, where the fluid permeates or is dissolved and held within the filter media. When adsorption is in play, the water, or other fluid to be filtered, passes through the carbon capturing contaminants and effectively reducing them.
Filters that employ activated carbon are rated by the size of the particles they can reduce, as measured in micron. This nominal micron rating can range from 50 micron down to as small as 0.5, more than adequate to filter sediment.
Greater Filtration with Integrated Membrane Pre-Activated Carbon Technology (“I.M.P.A.C.T.”)
Integrated Membrane Pre-Activated Carbon Technology (“I.M.P.A.C.T.”), used in a variety of 3M™ Water Filtration Products High Flow Series products, combines a pharmaceutical-grade microporous membrane and a pre-activated carbon block in a single cartridge to dramatically reduce pressure drop and practically eliminate the need for pre-filtration. The integrated membrane provides superior sediment holding capabilities while reducing Cryptosporidium and Giardia cysts¹and water-borne bacteria**. Pre-activation of the carbon media is not required.
*Tested and verified by manufacturer’s laboratory.
**As tested with E.coli ATCC (11229). Tested and verified by manufacturer’s laboratory
¹Based on the use of Cryptosporidium parvum oocysts.
²3M™ Water Filtration Products (-S) Replacement Filter Cartridges are NSF certified under high flow series commercial modular listing. Visit www.nsf.org for specific claims associated with each product.
³Surfaces that are in contact with water are FDA CFR-21 Compliant.