ContactUsLinkComponent
3M™ 1H Series Filter Housing
Catalog # 76031-822
Supplier:  3M Healthcare
3M™ 1H Series Filter Housing
Catalog # 76031-822
Supplier:  3M Healthcare
Supplier Number:  7000125598
Specifications
  • Cat. No.:
    76031-822
  • Model:
    4464310
Specifications
About this item
The 3M™ 1H1 series cartridge filter housing is comprised of a welded steel head and sump, steel pedestal, and 304 SS cartridge post.
  • Rugged, welded construction
  • Bolt-on style head
  • Internal baffle design accepts one, single length 9 ¾ inch filter cartridge
  • 1-inch NPTF connection
The 3M™ 1H1 series cartridge filter housing’s internal baffle design accepts one, single length 9 ¾ inch industrial filter cartridge. Its rugged, all-welded construction is suitable for filtration of industrial liquids.
Applicable filter cartridges: 3M™ Betafine™ DP series filters, 3M™ Betapure™ AU series filters, 3M™ Micro-Klean™ RB series filters.