ContactUsLinkComponent
3M™ Sediment Reduction Cartridge for Model BEV150 Reverse Osmosis System, Model CTG-BEV, 'S' TYPE, 5599705
3M™ Sediment Reduction Cartridge for Model BEV150 Reverse Osmosis System, Model CTG-BEV, 'S' TYPE, 5599705
Catalog # 76028-347
Supplier:  3M Healthcare
undefined
3M™ Sediment Reduction Cartridge for Model BEV150 Reverse Osmosis System, Model CTG-BEV, 'S' TYPE, 5599705
Catalog # 76028-347
Supplier:  3M Healthcare
Supplier Number:  7000050392
Specifications
  • Cat. No.:
    76028-347
  • Model:
    5599705
Specifications
About this item
Reverse Osmosis Replacement Cartridge allows throughput of up to 444 gal/2018.47 L per day with blending, dependent on system.
  • Some of the Pre-Filter Cartridges contain Integrated Pre-Activated Carbon Technology (IMPACT) media
Our Commercial Reverse Osmosis Replacement Cartridge available parts and accessories include element insertion and removal tools, an element hold down spring and a magnet assembly. The cartridges allow throughput dependent on system to up to 444 gal/2018.47 L per day with blending.