ContactUsLinkComponent
3M™ Commercial Reverse Osmosis Scale Reduction System for Coffee, Hot Tea & Espresso, Model TSR150-4
3M™ Commercial Reverse Osmosis Scale Reduction System for Coffee, Hot Tea & Espresso, Model TSR150-4
Catalog # 76031-302
Supplier:  3M Healthcare
undefined
3M™ Commercial Reverse Osmosis Scale Reduction System for Coffee, Hot Tea & Espresso, Model TSR150-4
Catalog # 76031-302
Supplier:  3M Healthcare
Supplier Number:  7010379420
Specifications
  • Description:
    Reverse Osmosis Scale Reduction System, 568 L Capacity
  • Cat. No.:
    76031-302
  • Model:
    TSR150-4
Specifications
About this item
3M™ water filtration products reverse osmosis water filtration system helps produce consistent Recipe Quality Water™ for hot beverages and steamers while maintaining efficient equipment operations in water conditions with high total dissolved solids (TDS) and/or hardness.
  • Can easily double RO production by adding RO and pre–filter expansion heads (sold separately)
  • Pre–Filter Cartridge Contains Integrated Pre–Activated Carbon Technology (I.M.P.A.CT.) media
  • Alerts the operator when the system requires maintenance
  • Allows store owner to dial in the desired level of minerals
  • The integrated heavy duty booster pump provides long pump life, helps reduce maintenance costs, and runs quietly
  • Real–time pressure and TDS monitoring
The unique design allows for filtered water to be blended with RO water for adjustment of beverage quality and production volumes to about 300 gal/day without blending or 444 gal/day with blending.